missing translation for 'onlineSavingsMsg'
Learn More

Vinylcyclopentane, 97%

Product Code. 10244904 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10244904 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10244904 Supplier Thermo Scientific Acros Supplier No. 150050010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3742-34-5 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3742-34-5
Color Colorless
Density 0.7000g/mL
Boiling Point 97.0°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H12
Linear Formula C5H9CH=CH2
Refractive Index 1.436
MDL Number MFCD00001386
Quantity 1 g
Specific Gravity 0.7
Synonym vinylcyclopentane, cyclopentane, ethenyl, cyclopentane, vinyl, cyclopentyl-ethene, cyclopentylethylene, cyclopentane,ethenyl, acmc-209iss
InChI Key BEFDCLMNVWHSGT-UHFFFAOYSA-N
SMILES C=CC1CCCC1
IUPAC Name ethenylcyclopentane
Molecular Weight (g/mol) 96.17
PubChem CID 77345
Formula Weight 96.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Vinylcyclopentane
Show More Show Less
compliance-icons
Product Identifier
  • Vinylcyclopentane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.