missing translation for 'onlineSavingsMsg'
Learn More

Vinylidene chloride, 99.9%, stabilized

Product Code. 10459300 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10459300 2.5 L Glass bottle 2.5 L
10449300 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10459300 Supplier Thermo Scientific Acros Supplier No. 172290025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 75-35-4 | C2H2Cl2 | 96.938 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-35-4 , 150-76-5
MDL Number MFCD00011653
InChI Key LGXVIGDEPROXKC-UHFFFAOYSA-N
Synonym vinylidene chloride, 1,1-dichloroethylene, vinylidene dichloride, 1,1-dce, ethene, 1,1-dichloro, vinylidine chloride, ethylene, 1,1-dichloro, as-dichloroethylene, rcra waste number u078, asym-dichloroethylene
PubChem CID 6366
ChEBI CHEBI:34031
IUPAC Name 1,1-dichloroethene
SMILES C=C(Cl)Cl

Specifications

Melting Point -122.0°C
Color Colorless
Density 1.2180g/mL
Boiling Point 31.2°C to 31.7°C
Flash Point −25°C
Infrared Spectrum Authentic
Assay Percent Range 99.85% min. (GC)
Packaging Glass bottle
Molecular Formula C2H2Cl2
Linear Formula H2C=CCl2
Refractive Index 1.4244 to 1.4264
Quantity 2.5 L
Beilstein 01, 186
Merck Index 15, 10192
Specific Gravity 1.218
Solubility Information Solubility in water: 2.5g/L (25°C). Other solubilities: soluble in most common organic solvents and,greases
Stabilizer 180 to 220ppm MEHQ
Molecular Weight (g/mol) 96.94
Viscosity .377 mPa.s (15°C)
Formula Weight 96.94
Percent Purity 99.9%
Physical Form Liquid
Chemical Name or Material Vinylidene chloride, 99.9%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Vinylidene chloride, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H224-Extremely flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.