missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Vinylidene chloride, 99%, stab. with 200 ppm 4-methoxyphenol

Product Code. 11373456
Change view
Click to view available options
Quantity:
100mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373456 100mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11373456 Supplier Alfa Aesar™ Supplier No. L14308.AE
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Applications
Vinylidene chloride is used as pharmaceutical intermediate and in chemical research.

Solubility
Soluble in water (2.5g/L at 20°C).

Notes
It is sensitive to light and moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Store under inert gas.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-35-4
Boiling Point 30°C to 32°C
Flash Point −25°C (−13°F)
Odor Aromatic
Molecular Formula C2H2Cl2
Linear Formula H2C=CCl2
MDL Number MFCD00011653
Quantity 100mL
UN Number UN1303
Beilstein 1733365
Merck Index 14,9996
Synonym vinylidene chloride, 1,1-dichloroethylene, vinylidene dichloride, 1,1-dce, ethene, 1,1-dichloro, vinylidine chloride, ethylene, 1,1-dichloro, as-dichloroethylene, rcra waste number u078, asym-dichloroethylene
InChI Key LGXVIGDEPROXKC-UHFFFAOYSA-N
SMILES C=C(Cl)Cl
IUPAC Name 1,1-dichloroethene
Molecular Weight (g/mol) 96.938
PubChem CID 6366
ChEBI CHEBI:34031
Formula Weight 96.94
Percent Purity 99%
Chemical Name or Material Vinylidene chloride, stabilized with 200ppm 4-methoxyphenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.