missing translation for 'onlineSavingsMsg'
Learn More

Vinylphosphonic acid, 90%, technical, Thermo Scientific Chemicals

Product Code. 15532293 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15532293 50 g 50g
15522293 10 g 10g
15542293 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15532293 Supplier Thermo Scientific Acros Supplier No. 300590500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1746-03-8
Molecular Formula C2H3O3P
Molecular Weight (g/mol) 106.02
MDL Number MFCD00043866
InChI Key ZTWTYVWXUKTLCP-UHFFFAOYSA-L
Synonym vinylphosphonic acid, phosphonic acid, ethenyl-, homopolymer, vinyl-phosphonicaci, ethenephosphonicacid, ethenylphosphonicacid, ethenyl-phosphonicaci, ethylenephosphonicacid, phosphonic acid, p-ethenyl-, homopolymer, acmc-1bvr6, poly vinylphosphonic acid
PubChem CID 168725
IUPAC Name ethenylphosphonic acid
SMILES [O-]P([O-])(=O)C=C

Specifications

Melting Point 36.0°C
Density 1.3700g/mL
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (Acid)
Linear Formula H2C=CHP(O)(OH)2
Quantity 50 g
Specific Gravity 1.37
Solubility Information Solubility in water: soluble.
Formula Weight 108.03
Percent Purity 90%
Grade Technical
Chemical Name or Material Vinylphosphonic acid
Show More Show Less
compliance-icons
Product Identifier
  • Vinylphosphonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.