missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Viral Production Cells 2.0

Product Code. 17391889 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 mL
6 x 1 mL
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17391889 1 mL Each
17661546 6 x 1 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17391889 Supplier Gibco™ Supplier No. A49784
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Viral Production Cells 2.0 are a clonal cell line derived from the HEK293F parental cell line and a core component of the AAV-MAX Helper-Free AAV Production System.

Gibco Viral Production Cells 2.0 are a clonal cell line derived from the HEK293F parental cell line and a core component of the AAV-MAX Helper-Free AAV Production System.

  • Adapted to the chemically defined Viral Production Medium and maintained in suspension culture in that medium
  • Highly transfectable using the AAV-MAX Transfection Kit for adeno-associated virus (AAV) production
  • Supplied frozen in a vial containing 1 mL of cells at 1 x 107 viable cells/mL in Viral Production Medium and 10% DMSO
  • Should be thawed directly into Viral Production Medium supplemented with 4 mM GlutaMAX Supplement (Cat. No. 35050038) for optimal performance
  • Clonal, 293F-derived, high-producer cell line
  • Absence of SV40 large T antigen
  • Optimized for high-density, suspension culture (>12 million cells/mL) in chemically defined medium
  • Enable production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
  • Robust scalability and passage stability
TRUSTED_SUSTAINABILITY

Specifications

Cell State Frozen
Concentration 1 x 107 cells/mL
Content And Storage 1 cryovial, store in vapor phase liquid nitrogen
Cell Line Viral Production Cells 2.0
Cell Type Kidney (Embryonic)
Culture Type Suspension Cell Culture
Form Frozen
Species Human
Format 1 Vial
For Use With (Application) AAV Production
Media Recommendation Gibco Viral Production Medium
No. of Cells 1 x 107 cells
Product Line AAV-MAX
Product Type Viral Production Cells
Quantity 1 mL
Shipping Condition Dry ice for domestic shipment; liquid nitrogen for international shipment
Source Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.