missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Viral Production Medium, AGT™

Product Code. 30283139 Shop All Gibco Products
Change view
Click to view available options
Reconstituted Volume:
100 L
1 L
50 L
Unit Size:
1 L
100 L
50 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Reconstituted Volume Unit Size
30283139 100 L 100 L
30282803 1 L 1 L
30282829 50 L 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30283139 Supplier Gibco™ Supplier No. A5147104
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco Viral Production Medium, AGT, is the dry-format version of CTS Viral Production Medium, a chemically defined, serum-free, protein-free, animal origin-free, phenol red-free medium.

  • Gibco Viral Production Medium, AGT is the dry-format version of CTS Viral Production Medium, a chemically defined, serum-free, protein-free, animal origin-free, phenol red-free medium
  • Medium is part of the CTS AAV-MAX Helper-Free AAV Production System for high-titer adeno-associated virus (AAV) production
  • The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade and GMP versions to enable seamless transition from research to commercial manufacturing
  • Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0, CTS AAV-MAX Transfection Kit, and CTS Viral-Plex Complexation Buffer.

Features of Viral Production Medium, AGT, include:

  • Supports growth and transfection of CTS Viral Production Cells 2.0 from shake flask to bioreactor scales
  • Helps enable production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
  • Simplifies transfer to manufacturing scale due to Advanced Granulation Technology (AGT) enabling auto-pH and simplified dissolution
  • Formulated with GlutaMAX Supplement, no further supplementation required
  • Manufactured in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to ISO 13485. Drug Master File and Regulatory Support File available upon request.

Advanced Granulation Technology

  • Advanced Granulation Technology (AGT) helps simplify medium preparation at large scale with fast rehydration
  • A just-add water protocol, and low dust generation, creates less pressure build-up during sterile filtration
  • For more information on AGT media preparation at large scale, see Hydration and scale-up of AGT medium in HyPerforma and imPULSE Single-Use Mixers
TRUSTED_SUSTAINABILITY

Specifications

Cell Line Viral Production Cells 2.0
Cell Type HEK 293 cells
Concentration 25.6 g/L
Form Powder (AGT)
Product Type AAV Production Medium
Sterility Non-sterile
With Additives Glucose, GlutaMAX
Without Additives No Phenol Red, No Antibiotics
For Use With (Application) AAV Production
Product Line AAV-MAX
Quantity 100 L
pH 6.9 to 7.5
Reconstituted Volume 100 L
Content And Storage 100 L Ropack Bucket, store at 2°C to 8°C protected from light
Culture Type Suspension Cell Culture
Expression System Mammalian
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.