missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Viral Production Medium, AGT™
Click to view available options
Reconstituted Volume:
100 L
1 L
50 L
Unit Size:
1 L
100 L
50 L
Description
- Gibco Viral Production Medium, AGT is the dry-format version of CTS Viral Production Medium, a chemically defined, serum-free, protein-free, animal origin-free, phenol red-free medium
- Medium is part of the CTS AAV-MAX Helper-Free AAV Production System for high-titer adeno-associated virus (AAV) production
- The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade and GMP versions to enable seamless transition from research to commercial manufacturing
- Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0, CTS AAV-MAX Transfection Kit, and CTS Viral-Plex Complexation Buffer.
Features of Viral Production Medium, AGT, include:
- Supports growth and transfection of CTS Viral Production Cells 2.0 from shake flask to bioreactor scales
- Helps enable production of high AAV titers, greater than 5 x 1010 vg/mL (unpurified)
- Simplifies transfer to manufacturing scale due to Advanced Granulation Technology (AGT) enabling auto-pH and simplified dissolution
- Formulated with GlutaMAX Supplement, no further supplementation required
- Manufactured in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to ISO 13485. Drug Master File and Regulatory Support File available upon request.
Advanced Granulation Technology
- Advanced Granulation Technology (AGT) helps simplify medium preparation at large scale with fast rehydration
- A just-add water protocol, and low dust generation, creates less pressure build-up during sterile filtration
- For more information on AGT media preparation at large scale, see Hydration and scale-up of AGT medium in HyPerforma and imPULSE Single-Use Mixers
Specifications
Specifications
| Cell Line | Viral Production Cells 2.0 |
| Cell Type | HEK 293 cells |
| Concentration | 25.6 g/L |
| Form | Powder (AGT) |
| Product Type | AAV Production Medium |
| Sterility | Non-sterile |
| With Additives | Glucose, GlutaMAX |
| Without Additives | No Phenol Red, No Antibiotics |
| For Use With (Application) | AAV Production |
| Product Line | AAV-MAX |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction