missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ViralSEQ™ dPCR AAV Titer Kit

Product Code. 18705515 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18705515 100 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18705515 Supplier Applied Biosystems™ Supplier No. A59364
  • ONLINE EXCLUSIVE PRICE
    1895.00 €
    1780.00 € / Pack of 1
    Save 115.00 €
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Applied Biosystems ViralSEQ dPCR AAV Titer Kit is a digital PCR (dPCR) assay for the reproducible quantitation of AAV particles independent of the serotype.

The Applied Biosystems ViralSEQ dPCR AAV Titer Kit is a digital PCR (dPCR) assay for the reproducible quantitation of AAV particles independent of the serotype. AAV titer quantitation is a critical quality attribute (CQA) in viral vector manufacturing for gene therapy development. This probe-based dPCR assay delivers superior sensitivity and consistency, outperforming conventional DNA blotting or ELISA methods.

Features of the ViralSEQ dPCR AAV Titer Kit include:

  • Accurate quantitation of viral particles by dPCR targeting ITR and SV40
  • Assay design compatible with more than 80% of AAV transfer plasmids
  • Easy-to-use, integrated sample-to-results system features TaqMan dPCR master mix and TaqMan primer/probe set
  • Highly sensitive quantitation delivers results in about three hours
  • Optional manual or automated sample preparation, optimized for quantitative recovery from bioproduction samples

The high sensitivity and specificity provided by the ViralSEQ dPCR AAV Titer Kit make it suitable for viral quantitation in process development and lot release testing.

Digital PCR workflow
The ViralSEQ dPCR AAV Titer Kit is part of an integrated dPCR workflow for viral vector characterization during biopharmaceutical manufacturing. Optional use of the Pharma KingFisher Apex 96 Deep-Well Magnetic Particle Processor with the PrepSEQ Nucleic Acid Sample Preparation Kit helps ensure high recovery of viral DNA with less labor and error than manual processing. The Pharma KingFisher Apex instrument can process up to 24 samples in triplicate, compared to three samples in triplicate using a manual method.

To help ensure performance that meets or exceeds regulatory compliance, the ViralSEQ kit has been internally validated on the QuantStudio Absolute Q Digital PCR System. Data analysis is streamlined using the QuantStudio Absolute Q dPCR Software, which includes accurate quantitation and security, audit, and e-signature capabilities to help enable 21 CFR Part 11 compliance.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 5X Absolute Q Digital PCR Master Mix, 0.36 mL; Store at -15°C to -25°C, protect from light
• dPCR AAV Titer Assay Mix, 90 μL; Store at -15°C to -25°C, protect from light
• Negative Control (water), 1 mL; Store at -15°C to -25°C before first use and 2°C to 8°C after first use
Detection Method Digital PCR
Form Frozen
PCR Method Digital PCR
Label or Dye FAM, VIC, ABY
Product Line ViralSEQ
Product Type dPCR AAV Titer Kit
Quantity 100 reactions
For Use With (Application) Virus Detection
No. of Reactions 100 Reactions

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.