Learn More
Applied Biosystems™ ViralSEQ™ dPCR Lentivirus Proviral Titer Kit
Shop All Applied Biosystems ProductsDescription
The Applied Biosystems ViralSEQ dPCR Lentivirus Proviral DNA Titer Kit provides a wide dynamic range and high sensitivity for accurate detection of integrated provirus copies. With the kit, proviral copy number can be used to calculate infectious lentiviral titers (as transducing units per mL (TU/mL)) and volume of recombinant lentivirus needed to transduce cells at the desired multiplicity of infection (MOI). The kit can also be used to determine vector copy number, a critical quality attribute (CQA) in cell and gene-modified cell therapy development. The assay targets a conserved region in the lentivirus genome and is designed to be compatible with most lentiviral transfer plasmids.
Features of the ViralSEQ dPCR Lentivirus Proviral DNA Titer Kit include:
- Accurate quantitation of integrated genome copies by targeting a conserved region in the lentivirus genome
- Assay design compatibility with more than 200 lentiviral transfer plasmids
- Easy-to-use, integrated sample-to-results system features TaqMan dPCR master mix and TaqMan primer/probe set
- Highly sensitive quantitation delivers results in about three hours
- Optional manual or automated sample preparation, optimized for quantitative recovery from bioproduction samples
The high sensitivity and specificity provided by the ViralSEQ dPCR Lentivirus Proviral DNA Titer Kit make it suitable for proviral quantitation in process development and lot release testing.
Digital PCR workflow
The ViralSEQ dPCR Lentivirus Proviral DNA Titer Kit is part of an integrated digital PCR (dPCR) workflow for viral vector characterization during biopharmaceutical manufacturing. Optional use of the Pharma KingFisher Apex 96 Deep-Well Magnetic Particle Processor with the PrepSEQ Nucleic Acid Sample Preparation Kit helps ensure high recovery of viral DNA with less labor and error than manual processing. The Pharma KingFisher Apex instrument can process up to 24 samples in triplicate, compared to three samples in triplicate using a manual method.
To help ensure performance that meets or exceeds regulatory compliance, the ViralSEQ kit has been internally validated on the QuantStudio Absolute Q Digital PCR System. Data analysis is streamlined using the QuantStudio Absolute Q dPCR Software, which includes accurate quantitation and security, audit, and e-signature capabilities to help enable 21 CFR Part 11 compliance.
Specifications
Specifications
| Content And Storage | • 5X Absolute Q Digital PCR Master Mix, 0.36 mL; Store at -15°C to -25°C, protect from light • dPCR Lentivirus Proviral Titer Assay Mix, 180 μL; Store at -15°C to -25°C, protect from light • Negative Control (water), 1 mL; Store at -15°C to -25°C before first use and 2°C to 8°C after first use |
| Detection Method | Digital PCR |
| PCR Method | Digital PCR |
| Label or Dye | FAM + VIC |
| Product Line | ViralSEQ |
| Product Type | dPCR Lentivirus Proviral Titer Kit |
| Quantity | 100 reactions |
| For Use With (Application) | Virus Detection |
| No. of Reactions | 100 Reactions |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.