Learn More
Invitrogen™ ViraPower™ HiPerform™ T-REx™ Gateway™ Vector Kit
Description
The ViraPower™ HiPerform™ T-REx™ Gateway™ Vector Kit contains the Gateway™-adapted ViraPower™ HiPerform™ tetracycline-regulated lentiviral expression vector, pLenti6.3/TO/V5-DEST™ for easy recombination-based cloning and lentiviral-based, regulated, high-level expression of a target gene in dividing and non-dividing mammalian cells. The pLenti6.3/TO/V5-DEST™ vector is equipped with two key genetic elements, making it a HiPerform™ vector: the Woodchuck Posttranscriptional Regulatory Element (WPRE) and the central Polypurine Tract (cPPT) sequence from the HIV-1 integrase gene to produce at least 4-fold increase in protein expression compared to vectors lacking these elements.
Advantages
- Generates replication-incompetent lentivirus for transducing dividing and non-dividing mammalian cells
- Easy recombination-based cloning using Gateway™ technology
- Stable, long-term, tetracycline-regulated expression
- Enhanced protein expression, up to 4-fold or greater, compared to traditional lentiviral expression systems
Key Features
- WPRE from the woodchuck hepatitis virus, increases transgene expression and cPPT from the HIV-1 integrase gene, increases the copy number of lentivirus integrating into the host genome, thus increasing viral titer. WPRE and cPPT together produce at least a four-fold increase in protein expression in most cell types, compared to other vectors that do not contain these elements.
- Hybrid promoter consisting of the human cytomegalovirus (CMV) promoter and two tetracycline operator 2 (TetO2) sites for high-level, regulated expression of the target gene
- Blasticidin selection marker for stable selection under control of SV40 promoter
Kit includes
- pLenti6.3/TO/V5-DEST™ vector
- pLenti3.3/TR vector
- pLenti6.3/TO/V5-GW/lacZ plasmid
- Tetracycline
- One Shot™ Stbl3™ Chemically Competent E. coli (Cat. No. C737303)
- pENTRTM Gus positive control plasmid.
Specifications
Specifications
| Constitutive or Inducible System | Inducible |
| Delivery Type | Lentiviral |
| Inducing Agent | Doxycycline, Tetracycline |
| Promoter | CMV/TO |
| Product Type | Vector Kit |
| Selection Agent (Eukaryotic) | Blasticidin |
| Content And Storage | ViraPower™ HiPerform™ T-REx™ Gateway™ Vector Kit • pLenti6.3/TO/V5-DEST™ vector (40 μL of vector at 150 ng/μL in TE Buffer, pH 8.0), 6 μg • pLenti3.3/TR vector (40 μL of vector at 0.5 μg/μL in TE Buffer, pH 8.0), 20 μg • pLenti6.3/TO/V5-GW/lacZ plasmid (20 μL of plasmid at 0.5 μg/μL in TE Buffer, pH 8.0), 10 μg • Tetracycline (10 mg/mL in water), 1 mL. Store at -20°C and protect from light. Store at -20°C unless otherwise noted One Shot™ Stbl3™ Chemically Competent E. coli (Cat. No. C737303) • Stbl3™ Cells in 6 mL of S.O.C. Medium, 21 x 50 μL • pUC19 control DNA (10 pg/μL), 50 μL Store at -80°C pENTR™ Gus positive control plasmid, store at -20°C. |
| Antibiotic Resistance Bacterial | Ampicillin (AmpR), Blasticidin (BsdR) |
| Cleavage | No Cleavage Site |
| Gateway Enzyme | LR Clonase™ II Plus, LR Clonase™ II |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.