missing translation for 'onlineSavingsMsg'
Learn More

Day Impex™ Virkon™ Virucidal Disinfectant

Product Code. 31759331
Change view
Click to view available options
Volume (Metric):
1000 mL
Unit Size:
4kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
31759331 1000 mL 4kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Read instructions before use or contact manufacturer for the latest Health and Safety advice
Product Code. 31759331 Supplier Day Impex™ Supplier No. 330006
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Read instructions before use or contact manufacturer for the latest Health and Safety advice

Clean and disinfect work surfaces and equipment in a single, one-step operation with this virucidal disinfectant.

  • Use biocides safely. Always read the label and product information before use
  • Widest proven spectrum of any disinfectant - proven effective in independent tests against all 18 virus families affecting man and animals, including AIDS (HIV) and Hepatitis B
  • Excellent bactericide and fungicide - cleans and disinfects work surfaces and equipment in a single, one-step operation
  • Can be applied in powder form direct to spillages of blood and other body fluids
  • Easily dissolves to make a liquid disinfectant
  • Odorless, non-irritant to skin and eyes, non-toxic vapor phase, exceptionally low oral toxicity
  • Under COSHH requirements, no maximum exposure limit is required
  • Non-corrosive, non-bleaching and biodegradable
  • Any viral disease transmitted via surfaces or instruments contaminated with infected blood, body fluids, feces or saliva can be controlled by correct application
  • For cleaning and disinfecting work surfaces and equipment

Specifications

Product Type Virucidal Disinfectant
Color-coded Yes
Form Granular Powder
Scent Lemon
Sterility Non-sterile
Container Type Tub
Quantity 4/Pk.
Volume (Metric) 1000 mL
compliance-icons
Product Identifier
  • RELYON VIRKON
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .

UK biocidal regulations (HSE / BPR)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.