missing translation for 'onlineSavingsMsg'
Learn More

Virkon™ Virucidal Disinfectant

Product Code. 12358667
Change view
Click to view available options
Volume (Metric):
50 g
500 g
5 kg
Container Type:
Drum
Sachet
Squeeze Pack
Quantity:
1 Ea.
50/Pk.
6/Pk.
Unit Size:
5kg
Pack of 50
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Container Type Quantity Unit Size
12358667 5 kg Drum 1 Ea. 5kg
12338667 50 g Sachet 50/Pk. Pack of 50
12348667 500 g Squeeze Pack 6/Pk. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Use biocides safely. Always read the label and product information before use
Product Code. 12358667 Supplier Virkon™ Supplier No. 330003
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Use biocides safely. Always read the label and product information before use

Clean and disinfect work surfaces and equipment in a single, one-step operation. Virkon™ Virucidal Disinfectant provides the widest proven spectrum of any disinfectant.

  • Use biocides safely. Always read the label and product information before use
  • Widest proven spectrum of any disinfectant - proven effective in independent tests against all 18 virus families affecting man and animals, including AIDS (HIV) and Hepatitis B
  • Excellent bactericide and fungicide - cleans and disinfects work surfaces and equipment in a single, one-step operation
  • Can be applied in powder form direct to spillages of blood and other body fluids
  • Easily dissolves to make a liquid disinfectant
  • Odourless, non-irritant to skin and eyes, non-toxic vapour phase, exceptionally low oral toxicity
  • Under COSHH requirements, no maximum exposure limit is required
  • Non-corrosive, non-bleaching and biodegradable
  • Any viral disease transmitted via surfaces or instruments contaminated with infected blood, body fluids, faeces or saliva can be controlled by correct application
  • For cleaning and disinfecting work surfaces and equipment

Specifications

Product Type Virucidal Disinfectant
Container Type Drum
For Use With (Application) Virucidal Disinfectant
Includes Measuring Beaker and Scoop
Quantity 1 Ea.
Volume (Metric) 5 kg
compliance-icons
Product Identifier
  • Rely+On Virkon
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains Dipotassium peroxodisulphate, Dipentene. May produce an allergic reaction.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.