missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ViroCheck NanoParticle Reference Kit

Product Code. 17636985
Change view
Click to view available options
Quantity:
25 tests kit
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17636985 25 tests kit Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17636985 Supplier Invitrogen™ Supplier No. V10425
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The ViroCheck NanoParticle Reference Kit is used for calibration of an intensity unit of fluorescence for flow cytometry analysis, and concentration analysis for virus and extracellular vesicles.

  • For calibration of an intensity unit of fluorescence for flow cytometry analysis and concentration analysis for virus and extracellular vesicles
  • Through the application of the NIST-assigned intensity standards for a given fluorophore, instrument sensitivity and accuracy can be determined
  • This permits data comparison between flow cytometry instruments
  • In combination with a biological standard, these particles can also be used to quantify the level of biomarker expression

ViroCheck NanoParticle Reference Kit features include:

  • NIST-assigned ERF values to 15 commonly used flow cytometry filter set channels
  • Enables normalization of the fluorescence intensity scale for instrument accuracy and sensitivity
  • NIST-verified nanoparticle concentration for counting of sub-micron biological samples such as viruses and extracellular vesicles (EV)
  • Facilitates instrument to instrument data comparison
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage
  • 100-nm FITC-like reference nanoparticles
  • 100-nm PE-like reference nanoparticles
  • 100-nm APC-like reference nanoparticles
  • 200-nm multicolor reference nanoparticles
  • 500-nm multicolor reference nanoparticles
For Use With (Application) Flow Cytometry
For Use With (Equipment) Flow Cytometers
No. of Tests 25 tests
Quantity 25 tests kit
Regulatory Status RUO
Shipping Condition 2° to 8°C
Stability Do Not Freeze
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.