Learn More
Poulten Selfe™ Viscometer Falling Sphere
Product Code. 13191003
Change view
Click to view available options
Product Type:
Falling Ball Viscometer
Unit Size:
Each
Product Code. Product Type Unit Size
13191003 Falling Ball Viscometer Each
1 options
Product Code. 13191003 Supplier Poulten Selfe™ Supplier No. 1715/03
  • ONLINE EXCLUSIVE PRICE
    145.00 €
    129.00 € / Pack of 1
    Save 16.00 €
    11% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For transparent liquids of high viscosity possessing Newtonian flow characteristics
For transparent liquids of high viscosity possessing Newtonian flow characteristics. Requires no calibrating liquid. Borosilicate glass 40mm diameter fall tube, graduated at six points, fitted by means of a 24–29 joint with a stopper having a sphere release tube with 5mm bore, funnel top and small vent hole. Without spheres.
Specifications
Product Type Falling Ball Viscometer