Learn More
Poulten Selfe™ Viscometer Holder
Product Code. 13171093
Change view
Click to view available options
Product Type:
U-tube Viscometer Holder
Ubbelohde Viscometer Holder
Size:
1 to 4
G, H
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Size Unit Size
13171093 U-tube Viscometer Holder G, H Each
13111103 Ubbelohde Viscometer Holder 1 to 4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13171093 Supplier Poulten Selfe™ Supplier No. 1818/A2
  • ONLINE EXCLUSIVE PRICE
    199.00 €
    196.82 € / Pack of 1
    Save 2.18 €
    1% Off
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Specifications
Product Type U-tube Viscometer Holder
For Use With (Equipment) U-tube Viscometer
For Use With (Application) 50 mm Bath Opening
Size G, H