Learn More
Poulten Selfe™ Viscometer Holder
Specifications
Specifications
| Product Type | Viscometer Holder |
| For Use With (Equipment) | Cannon-Fenske Routine 25 to 500 |
| Diameter (Metric) Opening | 50 mm |
| Size | Cannon-Fenske Size 25 to 500 |
Product Title
Haben Sie Verbesserungsvorschläge?