missing translation for 'onlineSavingsMsg'
Learn More

SCHOTT AG Lighting and Imaging™ VisiLED Extension Cable

Product Code. 30286565 Shop All Schott Products
Click to view available options
For Use With (Equipment):
VisiLED Controller MC-D 1100
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30286565

Brand: SCHOTT AG Lighting and Imaging™ 400031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optional 2.5 meter extension cable for connecting the lighting unit to the VisiLED controller MC-D 1100.

2.5 meter extension cable for connecting the VisiLED lighting unit to the VisiLED controller MC-D 1100.

Specifications

Type Cable
Product Line VisiLED
Autoclavable Not Autoclavable
Color Black
For Use With (Equipment) VisiLED Controller MC-D 1100
Length (Metric) 2.5 m
Sterility Non-sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.