Learn More
SCHOTT AG Lighting and Imaging™ VisiLED MC-D 1100
Product Code. 30286566 Shop All Schott Products
Change view
Click to view available options
Voltage:
100/240 V
Unit Size:
Each
Product Code. Voltage Unit Size
30286566 100/240 V Each
1 options
Product Code. 30286566 Supplier SCHOTT AG Lighting and Imaging™ Supplier No. 400060
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Digital controller for VisiLED microscopy illumination
  • With the VisiLED Controller MC-D 1100 you can seamlessly integrate our complete portfolio of VisiLED microscopy illumination into your own environments.
  • Designed for professional integration, the controller offers a robust solution to control our VisiLED light heads for stereo microscopy applications purely digital. T
  • he controller comes with a comprehensive Software Development Kit (SDK), featuring an extensive library of control commands.
  • Easy Integration into imaging and postprocessing software
  • Create tailor-made solutions according to unique customer needs
  • Complex lighting scenarios possible
  • Real-time control
  • Optimal contrasting thanks to broad VisiLED portfolio
  • Extensive control options
Specifications
Type LED Light Cube
Product Line VisiLED
Autoclavable Not Autoclavable
Dimensions (L x W x H) 160.7 x 110.7 x 32 mm
For Use With (Equipment) VisiLED Lightheads, Power supply unit and Power cord to be ordered separately
Voltage 100/240 V