missing translation for 'onlineSavingsMsg'
Learn More

PAL International™ White Visitors Lab Coats with Popper Fastening

Product Code. p-8003235
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12383996

Brand: PAL International™ N77113

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Offer hygienic protection in a range of industrial, food manufacturing and healthcare environments. PAL International™ White Visitors Lab Coats with Popper Fastening are made from soft but durable nonwoven polypropylene.

  • Soft but durable nonwoven polypropylene fabric construction
  • Strong and comfortable.
  • Open cuffs
  • No pockets
  • Individually bagged for each visitor
  • Food safe
  • Available in white only

Specifications

Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Open
Disposable Single-use
No. of Pockets None
For Use With (Application) Food Processing, Food Handling, Food Service
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.