missing translation for 'onlineSavingsMsg'
Learn More

Isofield™ Vista Cleanroom Sleeve Covers - Grade A/B Compatible

Product Code. 32016121
Change view
Click to view available options
Size:
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
32016121 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32016121 Supplier Isofield™ Supplier No. 80771
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These low-linting, chemically resistant sleeve covers feature thumb loops, static-dissipative performance and Class 1 cleanliness.

  • Covers are aseptically folded and provide extended arm protection for Grade A/B applications
  • Sterile cleanroom sleeve covers
  • Extra length 55 cm sleeve covers with thumb loops for full arm coverage
  • Aseptically folded
  • Class 1 for cleanliness
  • Static-dissipative
  • Tested against chemotherapy
  • Microporous polypropylene/polyethylene
  • 60 gsm, low-linting robust fabric

Technical and Documentation Compliance:

  • COA, COC, COI available for download online
  • QR code on cartons allows for immediate document retrieval
  • Type PB[6]
  • Resistance to penetration by spray EN 13034:2015 + A1:2009 / ISO 17491-4:2008
  • Inward leakage of aerosols and fine particles EN ISO 13982-2:2004 / EN ISO 13982-1:2004
  • Permeation of liquids and gases ASTM F739-20
  • Particulate cleanliness - Helmke Drum Test: Class 1
  • Surface resistivity ASTM D257
  • Sterility validation EN ISO 11137 Part 2:2015

Specifications

Size Universal
Construction Aseptically Folded
Product Type Sleeve Cover
Material Polyethylene, Non-woven Polypropylene Laminate
Color White
Sterility Sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Bound
Cuff Style Thumb Loop
Latex Free Yes
Model Vista
Thickness 2.6 mil
For Use With (Application) GMP Cleanroom, Pharmaceutical, Biotechnology, Medical Device, Laboratory, and Electronics Manufacturing Environments
Certifications/Compliance (Europe) PPE Regulation (EU) 2016/425
Includes Vista Cleanroom Sleeve Covers with Covered Elastic and Thumb Loops
Length (English) 21 in.
Length (Metric) 55 cm
No. per Case 100 Pairs
No. per Pack 10 Pairs
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.