missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies MLA™ Brand Selectable-Volume Macropipetter

Click to view available options
Volume (Metric):
2 to 5 mL
Unit Size:
Each
Description
Simple twist of bonnet permits selection of one of four factory-calibrated volumes: 2, 3, 4, and 5mL.
- Accurate to within ±1%†
- Uses disposable macropipetter tips (Mfr. No. 9048), which eliminate biohazard, breakage, and sterilization problems
Specifications
Specifications
| Accuracy | ±1% |
| Autoclavable | Not Autoclavable |
| Color | Blue |
| Disposable | Reusable |
| Grip Type | Ergonomic |
| Material | Aluminum, Stainless Steel |
| Number of Channels | 1 |
| Product Line | MLA Brand |
| Programmability | No |
| Sterility | Non-sterile |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction