missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Reagent Reservoirs
GREENER_CHOICE

Product Code. 11915732
Change view
Click to view available options
Capacity (Metric):
10 mL
100 mL
100 mL (with lid)
25 mL
25 mL (divided)
50 mL
Sterility:
Non-sterile
Sterile
Packaging:
Bulk
Individually Wrapped
Individually packed
Quantity:
10/bag x 10; 100/Cs.
100/Cs.
20/bag x 10; 200/Cs.
200 in. 20 bags of 10
50/Cs.
missing translation for 'unitSize'
Pack of 100
Pack of 200
Pack of 50
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Sterility Packaging Quantity unitSize
11915732 100 mL Sterile Bulk 10/bag x 10; 100/Cs. Pack of 100
11925732 100 mL Sterile Individually Wrapped 50/Cs. Pack of 50
11945732 25 mL (divided) Sterile Bulk 10/bag x 10; 100/Cs. Pack of 100
11925742 100 mL (with lid) Sterile Individually Wrapped 50/Cs. Pack of 50
11955732 25 mL Sterile Individually Wrapped 50/Cs. Pack of 50
11975732 50 mL Sterile Bulk 10/bag x 10; 100/Cs. Pack of 100
11985732 50 mL Sterile Individually Wrapped 50/Cs. Pack of 50
11935722 100 mL Sterile Individually Wrapped 100/Cs. Pack of 100
11995732 100 mL Sterile Bulk 20/bag x 10; 200/Cs. Pack of 200
11965722 50 mL Sterile Bulk 20/bag x 10; 200/Cs. Pack of 200
11945722 25 mL Sterile Bulk 20/bag x 10; 200/Cs. Pack of 200
11905742 50 mL Sterile Individually Wrapped 100/Cs. Pack of 100
11955722 25 mL Sterile Individually Wrapped 100/Cs. Pack of 100
11975742 25 mL Non-sterile Bulk 10/bag x 10; 100/Cs. Pack of 100
11985742 25 mL (divided) Non-sterile Bulk 10/bag x 10; 100/Cs. Pack of 100
11915752 50 mL Non-sterile Bulk 10/bag x 10; 100/Cs. Pack of 100
11965742 100 mL Non-sterile Bulk 10/bag x 10; 100/Cs. Pack of 100
11935732 25 mL Sterile Bulk 10/bag x 10; 100/Cs. Pack of 100
11965732 25 mL (divided) Sterile Individually Wrapped 50/Cs. Pack of 50
15742962 10 mL Sterile Bulk 100/Cs. Pack of 100
16445094 10 mL Sterile Individually packed 50/Cs. Pack of 50
16455094 10 mL Sterile Bulk 200 in. 20 bags of 10 Pack of 200
16465094 10 mL Sterile Individually packed 100/Cs. Pack of 100
16405104 10 mL Non-sterile Bulk 100/Cs. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11915732 missing translation for 'mfr' VistaLab™ Technologies missing translation for 'supplierNo' 30541000
Spotlight Offers
Use promo code 28036
View all promotions
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Unique trough-within-trough design maximizes reagent recovery

  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
  • 100mL model (Cat. No. 21-385-111) is supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile

Order Info

Reservoir lid sold separately (Cat. No. 21-385-110).

Specifications

Certifications/Compliance Pyrogen-free
Color White
Capacity (Metric) 100 mL
Dimensions (L x W x H) 5.72 x 3.36 x 1.27 in.
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
For Use With (Application) General Liquid Handling
Height (English) 1.27 in.
Length (English) 5.72 in.
Packaging Bulk
Quantity 10/bag x 10; 100/Cs.
Width (English) 3.36 in.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.