missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Reagent Reservoirs Accessory: Lid

Product Code. 11915742
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11915742 100 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11915742 Supplier VistaLab™ Technologies Supplier No. 30543000
  • ONLINE EXCLUSIVE PRICE
    187.00 €
    184.68 € / Pack of 100
    Save 2.32 €
    1% Off
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with 100mL reagent reservoirs

  • For 100mL reservoirs protects reagents from dust and minimize evaporation

Specifications

Certifications/Compliance RNase/DNase, Pyrogen-free
Color Clear
Capacity (Metric) 100 mL
Material Plastic
Sterility Sterile
Type Reservoir Lid
For Use With (Application) General Liquid Handling
Packaging Bulk
Quantity 100/Cs.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.