missing translation for 'onlineSavingsMsg'
Learn More

Vistusertib, MedChemExpress

Product Code. 16476686
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476686 100 mg 100mg
16426398 10 mg 10mg
16421176 200 mg 200mg
16418987 5 mg 5mg
16412507 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16476686 Supplier Medchem Express Supplier No. HY15247

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vistusertib (AZD2014) is an ATP competitive mTOR inhibitor with an IC50 of 2.81 nM. AZD2014 inhibits both mTORC1 and mTORC2 complexes.

Vistusertib (AZD2014) is an ATP competitive mTOR inhibitor with an IC50 of 2.81 nM. AZD2014 inhibits both mTORC1 and mTORC2 complexes.

Specifications

Chemical Name or Material Vistusertib
CAS 1009298-59-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H30N6O3
Quantity 100 mg
Synonym AZD2014
Solubility Information DMSO : ≥ 50 mg/mL (108.10 mM)
SMILES O=C(NC)C1=CC=CC(C2=CC=C3C(N=C(N4[C@@H](C)COCC4)N=C3N5[C@@H](C)COCC5)=N2)=C1
Molecular Weight (g/mol) 462.54
Formula Weight 462.54
Percent Purity 95.66%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.