missing translation for 'onlineSavingsMsg'
Learn More

Vitamin A acetate, MP Biomedicals™

Product Code. 11447481 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447481 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11447481 Supplier MP Biomedicals Supplier No. 0210325780
  • ONLINE EXCLUSIVE PRICE
    132.00 €
    124.95 € / 100g
    Save 7.05 €
    5% Off
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 127-47-9
Molecular Formula C22H32O2
Molecular Weight (g/mol) 328.496
MDL Number MFCD00019413
InChI Key QGNJRVVDBSJHIZ-QHLGVNSISA-N
Synonym retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650
PubChem CID 638034
ChEBI CHEBI:32095
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C

Specifications

Color Yellow
Quantity 100 g
Formula Weight 328.5
Physical Form Crystalline Powder
Chemical Name or Material Vitamin A acetate
compliance-icons compliance-icons
Product Identifier
  • Vitamin A Acetate
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H315-Causes skin irritation.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.