missing translation for 'onlineSavingsMsg'
Learn More

Vitamin A palmitate, 1.7 M.I.U./g

Product Code. 15485289
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485289 25 g 25g
15495289 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485289 Supplier Thermo Scientific Alfa Aesar Supplier No. J63022.14
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Retinyl palmitate is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. It is used as a dietary supplement as a lipophilic base for eg: soft gelatin capsule. | CAS: 79-81-2 | C36H60O2 | 524.874 g/mol

Retinyl palmitate is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. It is used as a dietary supplement as a lipophilic base for eg: soft gelatin capsule. Other applications include as pharmaceutical product which is used for manufacturing aqueous solutions using solubilizing agents such as cremophor. It is also used in the fortification of fatty foods such as margarine, spreads, oils, milk and dairy products. It is used in cosmetics for the preparation of creams and lotions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 79-81-2
Molecular Formula C36H60O2
Molecular Weight (g/mol) 524.874
MDL Number MFCD00019414
InChI Key VYGQUTWHTHXGQB-FFHKNEKCSA-N
Synonym retinyl palmitate, vitamin a palmitate, retinol palmitate, retinol, hexadecanoate, arovit, all-trans-retinyl palmitate, optovit-a, retinyl hexadecanoate, aquapalm, dispatabs tabs
PubChem CID 5280531
ChEBI CHEBI:17616
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC=C(C)C=CC=C(C)C=CC1=C(CCCC1(C)C)C

Spezifikation

Melting Point 28°C to 29°C
Packaging Glass bottle
Quantity 25 g
Beilstein 1917366
Solubility Information Soluble in chloroform,ether,and vegetable oils. Insoluble in water.
Formula Weight 524.87
Physical Form Low Melting Solid
Chemical Name or Material Vitamin A palmitate
compliance-icons
Produktidentifikator
  • Vitamin A Palmitate
Signalwort
  • Danger
Gefahrenkategorie
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Reproductive toxicity Category 1B
Gefahrenhinweis
  • H360-May damage fertility or the unborn child.
  • H413-May cause long lasting harmful effects to aquatic life.
Sicherheitshinweis
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Retinol

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.