missing translation for 'onlineSavingsMsg'
Learn More

Vitamin A palmitate, 1.7 M.I.U./g

Product Code. 15495289
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495289 100 g 100g
15485289 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495289 Supplier Thermo Scientific Alfa Aesar Supplier No. J63022.22
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
This item is not returnable. View return policy

Retinyl palmitate is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. It is used as a dietary supplement as a lipophilic base for eg: soft gelatin capsule. | CAS: 79-81-2 | C36H60O2 | 524.874 g/mol

Retinyl palmitate is a vitamin A analog that inhibits cell proliferation and induces cell differentiation. It is used as a dietary supplement as a lipophilic base for eg: soft gelatin capsule. Other applications include as pharmaceutical product which is used for manufacturing aqueous solutions using solubilizing agents such as cremophor. It is also used in the fortification of fatty foods such as margarine, spreads, oils, milk and dairy products. It is used in cosmetics for the preparation of creams and lotions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-81-2
Molecular Formula C36H60O2
Molecular Weight (g/mol) 524.874
MDL Number MFCD00019414
InChI Key VYGQUTWHTHXGQB-FFHKNEKCSA-N
Synonym retinyl palmitate, vitamin a palmitate, retinol palmitate, retinol, hexadecanoate, arovit, all-trans-retinyl palmitate, optovit-a, retinyl hexadecanoate, aquapalm, dispatabs tabs
PubChem CID 5280531
ChEBI CHEBI:17616
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC=C(C)C=CC=C(C)C=CC1=C(CCCC1(C)C)C

Specifications

Melting Point 28°C to 29°C
Packaging Glass bottle
Quantity 100 g
Beilstein 1917366
Solubility Information Soluble in chloroform,ether,and vegetable oils. Insoluble in water.
Formula Weight 524.87
Physical Form Low Melting Solid
Chemical Name or Material Vitamin A palmitate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.