missing translation for 'onlineSavingsMsg'
Learn More

Vitamin B12, MedChemExpress

Product Code. 16462096
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462096 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16462096 Supplier Medchem Express Supplier No. HYB0315
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Vitamin B12 is a vitamin. Vitamin B12 plays a key role in the normal functioning of the brain and nervous system, and for the formation of blood.

Vitamin B12 is a vitamin. Vitamin B12 plays a key role in the normal functioning of the brain and nervous system, and for the formation of blood.

Specifications

Chemical Name or Material Vitamin B12
CAS 68-19-9
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C63H88CoN14O14P
Quantity 500 mg
Synonym Cyanocobalamin
Solubility Information DMSO : 25 mg/mL (18.45 mM; Need ultrasonic) ∣H2O : 6.25 mg/mL (4.61 mM; ultrasonic and warming and heat to 60°C)
SMILES N#[C-][Co+3]123([N]4=CN(C(OC(CO)C5OP6([O-])=O)C5O)C7=C4C=C(C)C(C)=C7)[N]8=C9C(CCC(N)=O)C(CC(N)=O)(C)C8(C)C(C(CC(N)=O)C%10(CCC(NCC(C)O6)=O)C)[N-]1C%10=C(C)C(C(CCC(N)=O)C%11(C)C)=[N]2C%11=CC%12=[N]3C(C(CC(N)=O)(C)C%12CCC(N)=O)=C9C
Molecular Weight (g/mol) 1355.37
Formula Weight 1355.37
Percent Purity 99.68%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.