missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vitamin D2, 97+%

Product Code. 10279320 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10279320

Brand: Thermo Scientific Chemicals 422630050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vitamin D2, or Ergocalciferol, CAS # 50-14-6, a fat-soluble hormone that is important for calcium and phosphorus absorption and metabolism. | CAS: 50-14-6 | C28H44O | 396.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vitamin D2 (ergocalciferol) is a form of vitamin D, a fat-soluble hormone found in food sources
  • Vitamin D is important for calcium and phosphorus homeostasis, and can play a role in many biological processes including cell growth
  • Vitamin D2 has a lower binding affinity than Vitamin D3 for vitamin D binding protein

Applications

  • Research studies show that Vitamin D2 treatment in rats inhibits bladder tumor promotion
  • Studies show that the treatment of lung carcinoma cells with vitamin D2 in vitro resulted in increased ACE2 protein levels
  • Vitamin D2 has been seen to interfere with prion protein oligomerization in vitro
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-14-6
Molecular Formula C28H44O
Molecular Weight (g/mol) 396.66
MDL Number MFCD00166988,MFCD11656674
InChI Key MECHNRXZTMCUDQ-VLOQVYPSSA-N
Synonym ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina
PubChem CID 134129658
IUPAC Name (1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
SMILES CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C

Specifications

Absorbance 460 to 490 (265nm, EtOH 96%)
Melting Point 115.0°C to 119.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 97.0 to 103% (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 10215
Specific Rotation Condition +82.00 (25.00°C c=3,in acetone)
Specific Rotation + 82.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in common organic solvents
Formula Weight 396.65
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material Vitamin D2, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.