missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vitamin D2/D3 Competitive ELISA Kit

Product Code. 30829261
Click to view available options
Quantity:
96 Tests
This item is not returnable. View return policy

Product Code. 30829261

Brand: Invitrogen™ EEL150

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Vitamin D2 and vitamin D3 are two forms of vitamin D, a group of fat-soluble vitamins that play crucial roles in maintaining overall health. Vitamin D2, also known as ergocalciferol, is derived from plant sources. It is produced when ultraviolet (UV) light converts ergosterol, a compound found in certain fungi and plants, into vitamin D2. Vitamin D3, also called cholecalciferol, is synthesized in the skin of humans and animals upon exposure to UVB light from the sun. It is the most biologically active form of vitamin D and is also found in certain animal-based food sources. Both vitamin D2 and D3 undergo similar metabolic processes in the liver and kidneys to be converted into their active form, known as calcitriol. Calcitriol acts as a hormone that regulates calcium and phosphorus metabolism in the body.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 6.25 to 400 ng/mL
Assay Sensitivity 3.75 ng/mL
Conjugate Unlabeled
Product Type ELISA
Sample Type Cell Lysates, Plasma, Serum, Tissue Homogenates
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV <10
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 50 μL, Plasma 50 μL, Serum 50 μL, Tissue Homogenate 50 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Vitamin D2/D3
Test Time 1 hrs 20 mins
Total Assay Time 2 hrs 30 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.