missing translation for 'onlineSavingsMsg'
Learn More

Vitamin D2, MedChemExpress

Product Code. 16478208
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478208 10 g 10g
16450648 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478208 Supplier Medchem Express Supplier No. HY76542
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Vitamin D2 (Ergocalciferol), drived from plant sources or dietary supplements, could be used as supplement of Vitamin D.

Vitamin D2 (Ergocalciferol), drived from plant sources or dietary supplements, could be used as supplement of Vitamin D.

Specifications

Chemical Name or Material Vitamin D2
CAS 50-14-6
Color White
Purity Grade Notes Research
Molecular Formula C28H44O
Quantity 10 g
Synonym Ergocalciferol Calciferol Ercalciol
Solubility Information DMSO : 31.25 mg/mL (78.78 mM; Need ultrasonic)
SMILES C=C1/C(C[C@@H](O)CC1)=C\C=C2[C@@]3([H])[C@@](CCC\2)(C)[C@]([C@H](C)/C=C/[C@H](C)C(C)C)([H])CC3
Molecular Weight (g/mol) 396.65
Formula Weight 396.65
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.