missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vitamin D3, 99%, crystalline

Product Code. 10396590 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10396590 1 g Glass bottle 1g
10656164 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396590 Supplier Thermo Scientific Chemicals Supplier No. 140940010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Vitamin D3, 99%, CAS # 67-97-0, also known as cholecalciferol, is a steroid hormone produced in the skin or obtained from dietary sources. It is an essential mediator of bone calcium metabolism. | CAS: 67-97-0 | C27H44O | 384.648 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Vitamin D3 is a steroid hormone generated in the skin when the 7-dehydrocholesterol interacts with ultraviolet irradiation. It can also be found in several types of food for daily intake.

• Vitamin D3 binds to vitamin D receptors, thus modulating gene expression

Applications

• In in vivo studies, vitamin D3 has been implicated in the maintenance of blood calcium and phosphorus levels, bone metabolism, metabolic functions, and transcription regulation

• It modulates the proliferation and differentiation of both normal and cancer cells

• It presents in vitro antiproliferative and antimetastatic activities on breast, colon, and prostate cancer cells

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-97-0
Molecular Formula C27H44O
Molecular Weight (g/mol) 384.648
MDL Number MFCD00078131
InChI Key QYSXJUFSXHHAJI-YRZJJWOYSA-N
Synonym vitamin d3, cholecalciferol, calciol, colecalciferol, oleovitamin d3, arachitol, activated 7-dehydrocholesterol, vitamin d, colecalcipherol, colecalciferolum
PubChem CID 5280795
ChEBI CHEBI:28940
IUPAC Name (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
SMILES CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C

Specifications

Melting Point 82°C to 87°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 10216
Specific Rotation Condition +108.50 (25.00°C c=0.8,ethanol)
Specific Rotation +105° to +112° (25°C, 589nm) (c=0.8, ethanol) (1 dm, within 30 min.)
Formula Weight 384.64
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Vitamin D3
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Vitamin D3, crystalline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
  • H372-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.