Learn More
Thermo Scientific Chemicals Vitamin D3, 99%, crystalline
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Vitamin D3 is a steroid hormone generated in the skin when the 7-dehydrocholesterol interacts with ultraviolet irradiation. It can also be found in several types of food for daily intake.
• Vitamin D3 binds to vitamin D receptors, thus modulating gene expression
Applications
• In in vivo studies, vitamin D3 has been implicated in the maintenance of blood calcium and phosphorus levels, bone metabolism, metabolic functions, and transcription regulation
• It modulates the proliferation and differentiation of both normal and cancer cells
• It presents in vitro antiproliferative and antimetastatic activities on breast, colon, and prostate cancer cells
Specifications
Specifications
| Melting Point | 82°C to 87°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Merck Index | 15, 10216 |
| Specific Rotation Condition | +108.50 (25.00°C c=0.8,ethanol) |
| Specific Rotation | +105° to +112° (25°C, 589nm) (c=0.8, ethanol) (1 dm, within 30 min.) |
| Formula Weight | 384.64 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.