missing translation for 'onlineSavingsMsg'
Learn More

α-Vitamin E, MedChemExpress

Product Code. 16469126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469126 100 mg 100mg
16422549 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469126 Supplier Medchem Express Supplier No. HYN0683
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress α-Vitamin E ((+)-α-Tocopherol), a naturally occurring vitamin E form, is a potent antioxidant.

α-Vitamin E ((+)-α-Tocopherol), a naturally occurring vitamin E form, is a potent antioxidant.

Specifications

Chemical Name or Material α-Vitamin E
CAS 59-02-9
Color Colorless
Purity Grade Notes Research
Molecular Formula C29H50O2
Quantity 100 mg
Synonym (+)-α-Tocopherol D-α-Tocopherol
Solubility Information DMSO : 50 mg/mL (116.09 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES OC1=C(C)C(C)=C2C(CC[C@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)O2)=C1C
Molecular Weight (g/mol) 430.71
Formula Weight 430.71
Percent Purity 98.0%
Grade Research
Physical Form Viscous Liquid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.