missing translation for 'onlineSavingsMsg'
Learn More

Vitamin K1, TRC

Product Code. 17919994
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17919994 100 mg 100mg
17929534 10 mg 10mg
17927924 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17919994 Supplier Toronto Research Chemicals Supplier No. TRCV676080100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-80-0
Formula Weight 450.3498
InChI Formula InChI=1S/C31H46O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20+/t23-,24-/m1/s1
IUPAC Name 2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione
Molecular Formula C31 H46 O2
Molecular Weight (g/mol) 450.7
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC\C(=C\CC1=C(C)C(=O)c2ccccc2C1=O)\C
Synonym 1,4-Naphthalenedione, 2-methyl-3-(3,7,11,15-tetramethyl-2-hexadecenyl)-, [R-[R*,R*-(E)]]-, 1,4-Naphthalenedione, 2-methyl-3-[(2E,7R,11R)-3,7,11,15-tetramethyl-2-hexadecenyl]- (9CI), Phylloquinone (8CI), 2-Methyl-3-[(2E,7R,11R)-3,7,11,15-tetramethyl-2-hexadecen-1-yl]-1,4-naphthalenedione, 2-Methyl-3-phytyl-1,4-naphthoquinone, 2',3'-trans-Vitamin K1, Amri-K, Antihemorrhagic vitamin, AquaMEPHYTON, Combinal K1, Haemokion, Kanakion, Kativ N, Kaywan, Kenadion, Kephton, Kinadion, Konakion, Mephyton, Mono-Kay, Monodion, NSC 270681, Orakay, Pediatrico, Phylloquinone K1, Phytomenadione, Phytonadione, Phytylmenadione, Phytylmenaquinone, Reconval, Synthex P, Veda K1, Veta K1, Vitamin K1(20), trans-Phylloquinone, α-Phylloquinone
Chemical Name or Material Vitamin K1 (Phytonadione)

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.