Learn More
Emma Safety Footwear Vito (Frontier 114) Safety Shoes
The executive safety shoe that's made with the same care and attention to detail associated with high end luxury shoes. Brazilian 2-tone full grain leather combined with nubuck adds to the stylish look.
Brand: Emma Safety Footwear MM11409040
Additional Details : Weight : 1.50000kg
Description
- S3-Safety shoe with sole plate for wet work
- Thermoplastic Polyurethane (TPU) Outsole
- Full grain leather construction
- ESD certified
- Chrome-free lining
- Sanitized
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver & Wellness leather | |
40 |
Laces | |
Vito (Frontier 114) | |
Leather, Nubuck | |
Steel |