Learn More
Emma Safety Footwear Vito (Frontier 114) Safety Shoes
The executive safety shoe that's made with the same care and  attention to detail associated with  high end luxury shoes. Brazilian 2-tone full grain leather combined with nubuck adds to the stylish look.
Brand:  Emma Safety Footwear MM11409043
Additional Details : Weight : 1.50000kg
 View more versions of this product
Product Code. 15885771
  • 210.00 € / 1 pair
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Thermoplastic Polyurethane (TPU) Outsole
  • Full grain leather construction
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Black
Hydro-Tec™ Sanitized Silver & Wellness leather
43
Laces
Vito (Frontier 114)
Leather, Nubuck
Steel