missing translation for 'onlineSavingsMsg'
Learn More

ROBU™ by STEREON VitraPOR™ Porous Glass Beads for Cell Culture, Porosity 2

Product Code. 15185265
Change view
Click to view available options
Particle Size (Metric):
4.0 mm
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Particle Size (Metric) Unit Size
15185265 4.0 mm 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Handle small glass components with suitable eye and hand protection and avoid unnecessary abrasion or particle generation.
Product Code. 15185265 Supplier ROBU™ by STEREON Supplier No. 80068
  • ONLINE EXCLUSIVE PRICE
    649.00 €
    616.25 € / 1 L
    Save 32.75 €
    5% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Handle small glass components with suitable eye and hand protection and avoid unnecessary abrasion or particle generation.

Open-pore borosilicate glass 3.3 carriers for bioprocess development.

  • Nominal pore-size range of 40 -100 μm
  • Available in bead diameters of 1.5, 4.0 and 8.0 mm
  • Supplied in 1 L packs; selected variants may also be available in 250 mL

Specifications

Sterility Non-sterile
Product Type Microcarrier
Material Borosilicate Glass (3.3)
Particle Size (Metric) 4.0 mm

Suitability and biological performance depend on the cell line, medium, surface condition, reactor and process parameters. Validate the material in the intended process.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.