missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ VitroEase™ Apoferritin Standard

Product Code. 17681546 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 x 20 μL
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 17681546

Brand: Thermo Scientific™ A51362

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

The Thermo Scientific™ VitroEase™ Apoferritin Standard is a highly validated pure protein for use as a quality control sample for cryo-electron microscopy single particle analysis.

  • A highly validated pure protein for use as a quality control sample for cryo-electron microscopy single particle analysis
  • Designed specifically to meet the stringent needs for high protein purity, minimal protein aggregation, and intact protein structure
  • Protein sample grid preparation is a major bottleneck in cryo-electron microscopy (cryo-EM) single particle analysis (SPA), often requiring days or even weeks to optimize
  • Apoferritin, a 24-subunit protein (474 kDa), is considered the gold standard for cryo-EM SPA
  • Protein production is optimized and validated for use in cryo-EM grid preparation and analysis
  • High-quality, consistent protein standard documented via lot-specific certificates of analysis
  • Ready-to-use format with each single-use vial sufficient for 5 to 6 grid preparations
  • Standard is fit for use as a comparison for success for new users, or as quality control for expert users
TRUSTED_SUSTAINABILITY

Specifications

Concentration 3.5–4.0 mg/mL
For Use With (Application) Cryo-EM Single Particle Analysis (SPA)
Name Apoferrtin
Quantity 5 x 20 μL
Gene Alias ApoF
Gene Symbol FTH1, FTH
Sterility Non-sterile
Detection Method Cryo-EM
For Use With (Equipment) Cryo-Electron Microscopes, Mass spectrometers
Sample Type Purified protein
Technique MS, Cryo-EM SPA
Volume (Metric) 100 μL
Expression System Bacterial
Form Liquid
Color Colorless
Ligand Type Iron
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.