missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ VITsafe™ Wide Mouth Safety Wash Bottles
Description
Working with chemical substances that are sometimes dangerous requires a high level of responsibility and care. VITsafe™ safety wash-bottles fulfill all safety requirements for safe handling. Spray bottles are individually wrapped in clearly labeled polyethylene bags.
Constructed of durable, transparent materials
Constructed of durable, transparent materials
- Low-density polyethylene (LDPE) bottles
- Spray tubes made with autoclavable, chemically resistant polypropylene (PP)
- Wide-mouth bottles for easy and safe filling, even without a funnel
- Transparent material for easy identification and monitoring
- Material name in German, English, French and Spanish
- Chemical formula, CAS No, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
- In accordance to EC Directive No. 1272/2008 (GHS)
- Design eliminates almost all static overpressure
- Prevents virtually all leaking or dripping that can occur with temperature changes in the lab
- Color-coded caps support easy visual identification of the media
- Bottles utilize screw cap lids with GL threads for a strong, secure hold
- Finely drawn tip creates a precise spray jet for fine control
- Optimized medium backflow through the continuous spray tube
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Color | Transparent |
| Material | LDPE |
| Packaging | Individually wrapped in PE bag |
| Quantity | 12Pack |
| Screw Color | Yellow |
| Thread Size (Metric) | GL 45 |
| Mouth | Wide |
| Height (Metric) | 146 mm |
| Chemical Label | Isopropanol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?