missing translation for 'onlineSavingsMsg'
Learn More

AGMA VIVA Detergent

Product Code. 30066744
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
30066744 5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30066744 Supplier AGMA Supplier No. 4201
  • ONLINE EXCLUSIVE PRICE
    78.00 €
    70.82 € / Pack of 4
    Save 7.18 €
    9% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protect your public spaces with a concentrated general purpose detergent.

The maintenance products are designed to clean and protect hard surfaces quickly, easily, and economically, and are trusted by industry giants across the globe.

VIVA is a general-purpose specially formulated, fast acting, heavy duty maintenance cleaner. Unlike many competitive products, VIVA does not need rinsing with water, as a single wipe with the prepared solution will leave a bright surface free from streaks and suds. The product is particularly effective on oil and grease bound dirt.

  • Ideal for peripheral areas surrounding your Cleanrooms.
  • A general-purpose cleaner designed for ambulances, municipal authorities, halls, floors, walls and general machinery, indoors and out.
  • Delivery Method: apply via spray, mop, sponge, brush or cloth.
  • Description: fast-acting, heavy-duty maintenance cleaner.
  • Content: General-purpose detergent concentrate (pink solution)
  • Packaging Format: 4 x 5 L
  • Cleaning of components prior to assembly
  • Cleaning of tools and equipment
  • Preparation of surface for decal mounting
  • AGMA Isopropanol 99 wipes complies to Rolls-Royce CSS 269 (MCL104). AGMA Isopropanol 99 Wipes are listed on Rolls-Royce RRA 15721 NMD as a class A material (lab ref. A15934, valid until 01/03/2027).

Specifications

Product Type Detergent
Color-coded No
Form Liquid
Scent Unscented
Sterility Non-sterile
Appearance Clear
Autoclavable Not Autoclavable
Color Pink
Disposable Limited-use
Flammable No
For Use With (Application) Cleaning of peripheral areas to Cleanrooms e.g. uncontrolled areas
Volume (Metric) 5 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.