Learn More
Cytiva Vivaspin™ 20 Sample Concentrators
Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. Cytiva Life Vivaspin™ 20 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.
187.00 €
Specifications
Material (Membrane) | PES |
---|---|
Filtration Area | 6 cm2 |
Volume (Metric) | 5 to 20 mL |
Length (Metric) | 116 mm |
Width (Metric) | 30 mm |
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
10656375
|
Cytiva
28-9323-61 |
30 kDa |
187.00 €
Pack of 12 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
10219484
|
Cytiva
28-9323-58 |
3 kDa |
187.00 €
Pack of 12 |
In Stock
Log in to see stock. |
|||||
10646375
|
Cytiva
28-9323-59 |
5 kDa |
187.00 €
Pack of 12 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
10239294
|
Cytiva
28-9323-62 |
50 kDa |
187.00 €
Pack of 12 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
10774797
|
Cytiva
28-9323-63 |
100 kDa |
187.00 €
Pack of 12 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
10015804
|
Cytiva
28-9323-60 |
10 kDa |
187.00 €
Pack of 12 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.
- Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
- Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
- Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
- Compatible pH range: 1 to 9
- Active membrane area: 6.0cm2
For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
Specifications
PES | |
5 to 20 mL | |
30 mm |
6 cm2 | |
116 mm |