missing translation for 'onlineSavingsMsg'
Learn More

Vivid™ BOMCC Substrate

Product Code. 10696873
Change view
Click to view available options
Quantity:
0.1mg
1mg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10696873 1mg 1mg
11599266 0.1mg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10696873 Supplier Invitrogen Supplier No. P2980
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Vivid™ BOMCC Substrate

Vivid™ Substrates are blocked dyes that yield a minimal fluorescence signal until oxidation occurs. Cytochrome P450 metabolism at either of two potential oxidation sites releases the highly fluorescent product. They have superior fluorescence, solubility, and kinetic properties compared to conventional fluorogenic probes. This results in higher sensitivity, greater signal-to-noise ratio, and better assay reproducibility. When used in combination with CYP450 BACULOSOMES™ Plus Reagents as part of the Vivid™ CYP450 Screening Kits, the Vivid™ Substrates enable rapid, robust detection of cytochrome P450 inhibitors. This particular substrate (Vivid ™ BOMCC Substrate) yields a product that emits a blue fluorescence.

More About Vivid™ CYP450 Screening Kits
Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition. Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagent. CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme and human cytochrome P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.

>> View all Vivid™ CYP450 Screening Kits

Applications: high-throughput screening of enzyme-drug interactions, compound profiling for drug inhibition of cytochrome P450 isozymes, generation of predictive SAR models to guide compound acquisition

For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Vivid™ CYP450 Screening kits include Vivid™ CYP450 Fluorogenic substrates, BACULOSOMES™ CYP450 reagents, fluorescent standards, and an NADPH-regeneration system. Vivid™ CYP450 Fluorogenic substrates are also available separately. Store reaction buffers at +4°C or room temperature. Store BACULOSOMES™, the NADPH-regeneration system, and NADP+ at -80°C. Store the Vivid™ substrates and standards at +4°C, protected from light. (For long-term storage, resuspend in organic solvent and keep at -20°C.)
Detection Method Fluorescence
Product Line VIVID
Quantity 1mg
Substrate BOMCC
Substrate Properties Chemical Substrate
Substrate Type Cytochrome P450 Substrate
Shipping Condition Approved for shipment on Wet or Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.