missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Vivid™ Cyan Fluorescent Standard

Product Code. 10032952
Click to view available options
Quantity:
Each
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10032952

Brand: Thermo Scientific™ P2877

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vivid™ Fluorescent Standards represent the products formed during oxidation of Vivid™ Substrates by cytochrome P450 enzymes. These standards enable quantification of enzyme activity when used as part of the Vivid™ CYP450 Screening Kits.

Vivid™ Fluorescent Standards represent the products formed during oxidation of Vivid™ Substrates by cytochrome P450 enzymes. These standards enable quantification of enzyme activity when used as part of the Vivid™ CYP450 Screening Kits. Vivid™ Fluorescent Standards are included in the Vivid™ Screening Kits, but may also be purchased separately. They are supplied as a dried film.

More About Vivid™ CYP450 Screening Kits

Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition. Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagent. Vivid™ Fluorogenic Substrates are blocked dyes that yield minimal fluorescence signal until cleaved or hydroxylated. Oxidation at either of two potential sites releases the highly fluorescent product. CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme and human cytochrome P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.

Applications

  • High-throughput screening of enzyme-drug interactions
  • Compound profiling for drug inhibition of cytochrome P450 isozymes
  • Generation of predictive SAR models to guide compound acquisition
TRUSTED_SUSTAINABILITY

Specifications

Description Vivid™ Cyan Fluorescent St and ard, 1 Each, Powder, 0.1μM Concentration, 584.52 Molecular Weight, –20°C Storage Requirements, Used for Cell Viability, Proliferation and Function and Pharma and Biopharma and Protein Assays and Analysis a
Formulation Powder
Concentration 0.1 μm
Format Powder
For Use With (Application) Cell viability, proliferation and function and pharma and biopharma and protein assays and analysis and target-based adme/tox assays, protein biology, cell analysis, proliferation and function, cell metabolism, drug discovery and development, target-based
Quantity Each
Product Type Fluorescent Standard
Kit Contents Vivid™ CYP450 Fluorogenic substrates, BACULOSOMES™ CYP450 reagents, fluorescent standards and an NADPH-regeneration system. Vivid™ CYP450 Fluorogenic substrates are also available separately. Store reaction buffers at 4°C or room tem
Content And Storage • Vivid™ CYP450 Fluorogenic substrates
• BACULOSOMES™ CYP450 reagents
• Fluorescent standards
• NADPH-regeneration system.

Vivid™ CYP450 Fluorogenic substrates are also available separately.

Store reaction buffers at +4°C or room temperature. Store BACULOSOMES™, the NADPH-regeneration system, and NADP+ at -80°C. Store the Vivid™ substrates and standards at +4°C, protected from light. (For long-term storage, resuspend in organic solvent and keep at -20°C.)
Shipping Condition Wet Ice
Label or Dye Other Label(s) or Dye(s)
Product Line VIVID™
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.