missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vivid™ CYP1A2 Blue Screening Kit
Description
Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition. They provide an optimized method for studying isozyme-specific CYP450-drug interactions, metabolism, and inhibition. Vivid™ CYP450 Screening Kits offer:
Easy three-step procedure, 'mix and read' format, reactions at room temperature or 37°C
Superior fluorescent properties and kinetics compared to conventional fluorogenic probes
High signal-to-background ratio, broad dynamic range
Compatible with multiple assay formats from 96-well to 1536-well
Simple Mix and Read Kit Format
Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagents. The CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme (CYP1A2 in this case) and human cytochrome-P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.
Unique Vivid™ Reagents for Bright Fluorescent Signals and Low Background
Vivid™ Substrates are blocked dyes that yield minimal fluorescence signal until cleaved or hydroxylated. Oxidation at either of two potential sites releases the highly fluorescent product (Figure 1). They have superior fluorescence, solubility, and kinetic properties compared to conventional fluorogenic probes. This results in higher sensitivity, greater signal-to-noise ratio, and better assay reproducibility. Vivid™ EOMCC Substrate is the substrate included in this particular kit. This substrate yields a product that emits a blue fluorescence.
Flexible Assay Formats for Optimized Results
The sensitivity of the Vivid™ CYP450 assays allows detection of weak inhibitors and miniaturization to as little as 2μl per reaction. Assays may be set up in kinetic mode or in end-point mode to facilitate multi-plate screening (Figure 2). Assays may be performed at room temperature or 37°C.
Applications: high-throughput screening of enzyme-drug interactions, compound profiling for drug inhibition of cytochrome P450 isozymes, generation of predictive SAR models to guide compound acquisition
For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
Simple Mix and Read Kit Format
Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagents. The CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme (CYP1A2 in this case) and human cytochrome-P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.
Unique Vivid™ Reagents for Bright Fluorescent Signals and Low Background
Vivid™ Substrates are blocked dyes that yield minimal fluorescence signal until cleaved or hydroxylated. Oxidation at either of two potential sites releases the highly fluorescent product (Figure 1). They have superior fluorescence, solubility, and kinetic properties compared to conventional fluorogenic probes. This results in higher sensitivity, greater signal-to-noise ratio, and better assay reproducibility. Vivid™ EOMCC Substrate is the substrate included in this particular kit. This substrate yields a product that emits a blue fluorescence.
Flexible Assay Formats for Optimized Results
The sensitivity of the Vivid™ CYP450 assays allows detection of weak inhibitors and miniaturization to as little as 2μl per reaction. Assays may be set up in kinetic mode or in end-point mode to facilitate multi-plate screening (Figure 2). Assays may be performed at room temperature or 37°C.
Applications: high-throughput screening of enzyme-drug interactions, compound profiling for drug inhibition of cytochrome P450 isozymes, generation of predictive SAR models to guide compound acquisition
For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
Specifications
Specifications
| For Use With (Equipment) | Microplate Reader |
| Product Line | VIVID |
| Content And Storage | Vivid™ CYP450 Screening kits include Vivid™ CYP450 Fluorogenic substrates, BACULOSOMES™ CYP450 reagents, fluorescent standards, and an NADPH-regeneration system. Vivid™ CYP450 Fluorogenic substrates are also available separately. Store reaction buffers at +4°C or room temperature. Store BACULOSOMES™, the NADPH-regeneration system, and NADP+ at -80°C. Store the Vivid™ substrates and standards at +4°C, protected from light. (For long-term storage, resuspend in organic solvent and keep at -20°C.) |
| Detection Method | Fluorescence |
| Label or Dye | VIVID Blue |
| Product Type | Screening Kit |
| Quantity | >300 Assays |
| Format | 96-well Plate |
| Color | Blue |
| Shipping Condition | Dry Ice |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?