missing translation for 'onlineSavingsMsg'
Learn More

Vivid™ CYP2C9 Blue Screening Kit

Product Code. 10145052
Change view
Click to view available options
Quantity:
>300 assay(s)
Unit Size:
Each
Product Code. Quantity unitSize
10145052 >300 assay(s) Each
1 options
This item is not returnable. View return policy

Product Code. 10145052

Brand: Invitrogen P2861

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

For Use With (Equipment) Microplate Reader
Product Line VIVID
Content And Storage Vivid™ CYP450 Screening kits include Vivid™ CYP450 Fluorogenic substrates, BACULOSOMES™ CYP450 reagents, fluorescent standards, and an NADPH-regeneration system. Vivid™ CYP450 Fluorogenic substrates are also available separately. Store reaction buffers at +4°C or room temperature. Store BACULOSOMES™, the NADPH-regeneration system, and NADP+ at -80°C. Store the Vivid™ substrates and standards at +4°C, protected from light. (For long-term storage, resuspend in organic solvent and keep at -20°C.)
Detection Method Fluorescence
Label or Dye VIVID Blue
Product Type Screening Kit
Quantity >300 assay(s)
Format 96-well Plate
Color Blue
Shipping Condition Dry Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.