missing translation for 'onlineSavingsMsg'
Learn More

VLX600, TRC

Product Code. 30457117
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457117 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457117 Supplier Toronto Research Chemicals Supplier No. TRCV757175100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 1-(2-Pyridinyl)ethanone 2-(6-Methyl-5H-1,2,4-triazino[5,6-b]indol-3-yl)hydrazone
CAS 327031-55-0
Molecular Formula C17H15N7
Quantity 100 mg
Synonym 1-(2-Pyridinyl)ethanone 2-(6-Methyl-5H-1,2,4-triazino[5,6-b]indol-3-yl)hydrazone, VLX600
InChI Formula InChI=1S/C17H15N7/c1-10-6-5-7-12-14(10)19-16-15(12)22-24-17(20-16)23-21-11(2)13-8-3-4-9-18-13/h3-9H,1-2H3,(H2,19,20,23,24)/b21-11+
SMILES C\C(=N/Nc1nnc2c3cccc(C)c3[nH]c2n1)\c4ccccn4
IUPAC Name 6-methyl-N-[(E)-1-pyridin-2-ylethylideneamino]-5H-[1,2,4]triazino[5,6-b]indol-3-amine
Molecular Weight (g/mol) 317.35
Formula Weight 317.1389
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.