missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Volume Readout Petro for Buerkle™ Electric Barrel Pump Petro
Shop All Bürkle Products
Click to view available options
For Use With:
Electric Barrel Pump
Unit Size:
Each
Description
- For use with Buerkle™ Electric Barrel Pump Petro
- With cumulative total and suitable for calibration
- Supplied uncalibrated
Specifications
Specifications
| Color | Black |
| For Use With | Electric Barrel Pump |
| Type | Volume Readout Petro |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction