missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VU 0134992

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
VU 0134992 is a Kir4.1 potassium channel blocker (IC50 = 0.97 μM). Displays 10-fold selectivity for Kir4.1 over Kir4.1/5.1 concatemeric channels and >30-fold selectivity over Kir1.1, 2.1 and 2.2. Increases urine output and urinary Na+ and K+ excretion in volume-loaded rats.
Specifications
Specifications
| Quantity | 10 mg |
| Molecular Formula | C20H31BrN2O2 |
| Formula Weight | 411.38 |
| Product Type | Blocker |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction