missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VU 0364739 hydrochloride
GREENER_CHOICE

Product Code. 18036032
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18036032

Brand: Tocris Bioscience™ 4171/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PLD2 inhibitor

Potent and selective phospholipase D2 (PLD2) inhibitor (IC50 = 20 nM); displays 75-fold selectivity against PLD1 (IC50 = 1500 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1244640-48-9
Molecular Formula C26H28ClFN4O2

Specifications

Quantity 10 mg
Formula Weight 482.98
Purity >98%
Molecular Weight (g/mol) 482.984
Chemical Name or Material VU 0364739 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.